Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 8 | 1574 | ||
NM_002266.3 | NP_002257.1 | 8 | 1216 | |||
GenBank mRNA | AK024833.1 | - | 8 | 1084 | ||
AK058111.1 | - | 8 | 1085 | |||
AK291954.1 | - | 8 | 1083 | |||
BC005978.1 | - | 8 | 1083 | 4105058 | ||
BC013257.1 | - | 8 | 1079 | |||
BC034443.1 | - | 8 | 1042 | |||
BC053343.1 | - | 8 | 1072 | 4450470 | ||
BC067848.1 | - | 8 | 1315 | 4825235 | ||
BC146905.1 | - | 8 | 1070 | 9056833 | ||
BT006665.1 | - | 7 | 954 | |||
U09559.1 | - | 8 | 1086 | |||
U28386.1 | - | 8 | 1086 |