Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 8 | 1691 | ||
NM_002266.3 | NP_002257.1 | 8 | 1333 | |||
GenBank mRNA | AK024833.1 | - | 8 | 1201 | ||
AK058111.1 | - | 8 | 1202 | |||
AK291954.1 | - | 8 | 1200 | |||
BC005978.1 | - | 8 | 1200 | 4105058 | ||
BC013257.1 | - | 8 | 1196 | |||
BC034443.1 | - | 8 | 1159 | |||
BC053343.1 | - | 8 | 1189 | 4450470 | ||
BC067848.1 | - | 8 | 1432 | 4825235 | ||
BC146905.1 | - | 8 | 1187 | 9056833 | ||
BT006665.1 | - | 7 | 1071 | |||
U09559.1 | - | 8 | 1203 | |||
U28386.1 | - | 8 | 1203 |
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 9 | 1982 | |||
GenBank mRNA | AK055342.1 | - | 8 | 1853 |