Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004466.5 | NP_004457.1 | 3 | 1423 | ||
XM_011521054.2 | XP_011519356.1 | 3 | 1355 | |||
XM_011521055.2 | XP_011519357.1 | 3 | 1355 | |||
XM_011521056.2 | XP_011519358.1 | 3 | 1355 | |||
XM_011521057.2 | XP_011519359.1 | 3 | 1355 | |||
XM_011521058.2 | XP_011519360.1 | 3 | 1355 | |||
XM_011521059.2 | XP_011519361.1 | 3 | 1355 | |||
XM_011521060.2 | XP_011519362.1 | 3 | 1355 | |||
XM_017020435.1 | XP_016875924.1 | 3 | 1355 | |||
XM_017020436.1 | XP_016875925.1 | 3 | 1355 | |||
XM_017020437.1 | XP_016875926.1 | 3 | 1355 | |||
GenBank mRNA | AF001462.1 | - | 3 | 1076 | ||
AK312815.1 | - | 4 | 1383 | |||
BC039730.1 | - | 4 | 1385 | 5744533 | ||
U66033.1 | - | 3 | 1009 |