Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 26 | 3266 | ||
XM_005251930.1 | XP_005251987.1 | 27 | 3365 | |||
XM_005251931.1 | XP_005251988.1 | 26 | 3284 | |||
XM_005251932.1 | XP_005251989.1 | 25 | 3196 | |||
GenBank mRNA | AY823636.1 | - | 4 | 828 | ||
BC014188.2 | - | 14 | 2283 | |||
BC069268.1 | - | 26 | 3267 | 6340972 | ||
BX647920.1 | - | 8 | 1222 |