Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004958.3 | NP_004949.1 | 6 | 919 | ||
XM_005263438.2 | XP_005263495.1 | 6 | 926 | |||
XM_011541166.2 | XP_011539468.1 | 6 | 919 | |||
XM_017000900.1 | XP_016856389.1 | 3 | 382 | |||
XM_017000901.1 | XP_016856390.1 | 5 | 779 | |||
XM_017000902.1 | XP_016856391.1 | 6 | 919 | |||
GenBank mRNA | AB209995.1 | - | 6 | 900 | ||
AK302863.1 | - | 6 | 911 | |||
AK304273.1 | - | 6 | 908 | |||
BC117166.1 | - | 5 | 816 | 40125717 | ||
L34075.1 | - | 6 | 877 | |||
U88966.1 | - | 6 | 877 |