Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004958.3 | NP_004949.1 | 15 | 2502 | ||
XM_005263438.2 | XP_005263495.1 | 15 | 2509 | |||
XM_011541166.2 | XP_011539468.1 | 15 | 2502 | |||
XM_017000900.1 | XP_016856389.1 | 12 | 1965 | |||
XM_017000901.1 | XP_016856390.1 | 13 | 2253 | |||
XM_017000902.1 | XP_016856391.1 | 15 | 2502 | |||
GenBank mRNA | AB209995.1 | - | 15 | 2483 | ||
AK302863.1 | - | 15 | 2494 | |||
AK304273.1 | - | 15 | 2491 | |||
BC117166.1 | - | 14 | 2399 | 40125717 | ||
L34075.1 | - | 15 | 2460 | |||
U88966.1 | - | 15 | 2460 |