Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098204.1 | NP_001091674.1 | 4 | 666 | ||
NM_001098205.1 | NP_001091675.1 | 4 | 706 | |||
NM_001098206.1 | NP_001091676.1 | 4 | 683 | |||
NM_001098207.1 | NP_001091677.1 | 4 | 710 | |||
NM_001098208.1 | NP_001091678.1 | 3 | 789 | |||
NM_004966.3 | NP_004957.1 | 4 | 667 | |||
GenBank mRNA | AK001364.1 | - | 4 | 704 | ||
AK296696.1 | - | 5 | 487 | |||
AL542769.3 | - | 4 | 626 | |||
BC001432.2 | - | 3 | 461 | 3138435 | ||
BC004254.1 | - | 4 | 625 | 3633625 | ||
BC015580.2 | - | 4 | 582 | 4636807 | ||
BC016736.1 | - | 4 | 655 | 4104978 | ||
BC106008.1 | - | 4 | 594 | 6156823 | ||
L28010.1 | - | 1 | 330 |