Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004210.4 | NP_004201.3 | 6 | 3293 | ||
XM_005270269.3 | XP_005270326.1 | 6 | 2962 | |||
XM_005270270.3 | XP_005270327.1 | 6 | 2939 | |||
XM_011540331.2 | XP_011538633.1 | 7 | 3495 | |||
XM_011540332.2 | XP_011538634.1 | 7 | 3142 | |||
XM_011540333.2 | XP_011538635.1 | 6 | 3028 | |||
XM_017016909.1 | XP_016872398.1 | 6 | 2922 | |||
GenBank mRNA | AF029729.1 | - | 6 | 2944 | ||
BC026336.1 | - | 6 | 2980 | 4812302 |