Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042729.1 | NP_001036194.1 | 7 | 789 | ||
NM_001042747.1 | NP_001036212.1 | 7 | 848 | |||
NM_005248.2 | NP_005239.1 | 7 | 805 | |||
XM_006710452.2 | XP_006710515.1 | 8 | 856 | |||
XM_011541010.1 | XP_011539312.1 | 7 | 770 | |||
XM_011541011.2 | XP_011539313.1 | 5 | 569 | |||
XM_011541012.1 | XP_011539314.1 | 7 | 805 | |||
XM_011541013.2 | XP_011539315.1 | 7 | 805 | |||
XM_011541014.2 | XP_011539316.1 | 3 | 387 | |||
XM_017000673.1 | XP_016856162.1 | 5 | 425 | |||
XM_017000674.1 | XP_016856163.1 | 4 | 360 | |||
GenBank mRNA | AK130188.1 | - | 7 | 767 | ||
AK301078.1 | - | 5 | 563 | |||
BC064382.1 | - | 7 | 718 | 5734966 | ||
M19722.1 | - | 5 | 724 |