Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005253.3 | NP_005244.1 | 4 | 1654 | ||
XM_006711976.2 | XP_006712039.1 | 4 | 1858 | |||
XM_006711977.3 | XP_006712040.1 | 4 | 2711 | |||
XM_017003737.1 | XP_016859226.1 | 4 | 2367 | |||
GenBank mRNA | AK055579.1 | - | 4 | 1708 | ||
AK302622.1 | - | 4 | 808 | |||
AK315415.1 | - | 4 | 958 | |||
BC022791.1 | - | 4 | 1078 | 4637727 | ||
BX647822.1 | - | 4 | 830 | |||
CR542262.1 | - | 4 | 845 | |||
X16706.1 | - | 4 | 848 |