Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005261.3 | NP_005252.1 | 5 | 1256 | ||
NM_181702.2 | NP_859053.1 | 5 | 1141 | |||
XM_017013315.1 | XP_016868804.1 | 5 | 1460 | |||
GenBank mRNA | BC022010.2 | - | 5 | 1141 | 4823985 | |
U10550.1 | - | 5 | 1230 |