Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318900.1 | NP_001305829.1 | 13 | 2422 | ||
NM_001318901.1 | NP_001305830.1 | 16 | 2775 | |||
NM_001318902.1 | NP_001305831.1 | 15 | 2702 | |||
NM_001318904.1 | NP_001305833.1 | 14 | 3217 | |||
NM_001318905.1 | NP_001305834.1 | 16 | 3343 | |||
NM_001318906.1 | NP_001305835.1 | 14 | 3050 | |||
NM_005271.4 | NP_005262.1 | 13 | 2990 | |||
GenBank mRNA | AB209406.1 | - | 12 | 5290 | ||
AK094782.1 | - | 15 | 2654 | |||
AK122685.1 | - | 16 | 2758 | |||
BC112946.1 | - | 13 | 2773 | 6138490 | ||
J03248.1 | - | 13 | 2775 | |||
M20867.1 | - | 13 | 2809 | |||
M37154.1 | - | 13 | 2725 | |||
X07674.1 | - | 13 | 2725 | |||
X07769.1 | - | 13 | 2725 |