Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005273.3 | NP_005264.2 | 8 | 943 | ||
GenBank mRNA | AF501883.1 | - | 7 | 670 | ||
AK056750.1 | - | 3 | 1065 | |||
AK291592.1 | - | 8 | 818 | |||
AL535249.3 | - | 8 | 938 | |||
BC010073.2 | - | 8 | 867 | 4300639 | ||
BC012348.1 | - | 8 | 928 | 4556119 | ||
BC068003.1 | - | 8 | 853 | 6055863 | ||
CR541729.1 | - | 7 | 670 | |||
M16538.1 | - | 8 | 817 | |||
M36429.1 | - | 7 | 727 |