Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 11 | 2855 | ||
XM_005249015.3 | XP_005249072.1 | 12 | 2318 | |||
GenBank mRNA | AK026391.1 | - | 11 | 1517 | ||
AK293596.1 | - | 5 | 808 | |||
AK298042.1 | - | 11 | 1694 | |||
AK301738.1 | - | 10 | 1250 | |||
AK302480.1 | - | 10 | 1583 | |||
AK316391.1 | - | 11 | 1456 | |||
BC013959.2 | - | 11 | 1504 | 2961417 | ||
BC018366.1 | - | 11 | 2435 | 3924808 | ||
BC063657.1 | - | 8 | 1087 | |||
BT006648.1 | - | 11 | 1464 | |||
L25665.1 | - | 11 | 1414 |