Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098200.1 | NP_001091670.1 | 2 | 612 | ||
NM_005292.3 | NP_005283.1 | 3 | 804 | |||
XM_006719946.3 | XP_006720009.1 | 3 | 1614 | |||
GenBank mRNA | AF339781.1 | - | 2 | 560 | ||
BC008569.1 | - | 2 | 571 | 4338810 | ||
BC050646.1 | - | 3 | 750 | 4341573 | ||
BC066927.1 | - | 3 | 860 | 4373724 | ||
BI770379.1 | - | 2 | 611 | |||
BT009907.1 | - | 1 | 303 | |||
JQ308176.1 | - | 1 | 303 |