Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005294.2 | NP_005285.1 | 2 | 1373 | ||
XM_005251933.3 | XP_005251990.1 | 2 | 7580 | |||
GenBank mRNA | AK299686.1 | - | 3 | 1343 | ||
AK313539.1 | - | 2 | 1347 | |||
BC066885.1 | - | 1 | 877 | 6971731 | ||
BC066886.1 | - | 1 | 877 | 6971734 | ||
BC067532.1 | - | 1 | 877 | 6971732 | ||
BC067533.1 | - | 1 | 877 | 6971733 | ||
BC067534.1 | - | 1 | 877 | 6971735 |
Set Membership: |
G-protein coupled receptor |