Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004229.3 | NP_004220.2 | 31 | 4888 | ||
XM_005272701.2 | XP_005272758.1 | 31 | 4798 | |||
XM_011544000.2 | XP_011542302.1 | 32 | 4770 | |||
XM_017029962.1 | XP_016885451.1 | 32 | 4763 | |||
GenBank mRNA | AB006651.1 | - | 31 | 4867 | ||
AF135802.1 | - | 31 | 4820 | |||
AF304448.1 | - | 31 | 4867 | |||
AK023368.1 | - | 16 | 2781 | |||
BC132672.1 | - | 31 | 4859 | 40146694 | ||
BC132674.1 | - | 31 | 4857 | 40146696 |