Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005334.2 | NP_005325.2 | 23 | 6117 | ||
XM_006724815.2 | XP_006724878.1 | 23 | 6717 | |||
XM_006724816.2 | XP_006724879.1 | 23 | 6714 | |||
XM_011531147.2 | XP_011529449.1 | 23 | 6594 | |||
XM_011531148.2 | XP_011529450.1 | 23 | 6591 | |||
XM_017029471.1 | XP_016884960.1 | 22 | 6516 | |||
XM_017029472.1 | XP_016884961.1 | 18 | 4902 | |||
GenBank mRNA | L20010.1 | - | 23 | 5933 |