Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243900.2 | NP_001230829.1 | 6 | 790 | ||
NM_001320965.1 | NP_001307894.1 | 5 | 724 | |||
NM_001320966.1 | NP_001307895.1 | 5 | 582 | |||
NM_001320967.1 | NP_001307896.1 | 5 | 659 | |||
NM_005336.5 | NP_005327.1 | 5 | 806 | |||
NM_203346.4 | NP_976221.1 | 5 | 659 | |||
XM_005247002.3 | XP_005247059.2 | 5 | 613 | |||
XM_005247003.4 | XP_005247060.2 | 5 | 658 | |||
XM_006712475.3 | XP_006712538.1 | 7 | 862 | |||
XM_011511058.2 | XP_011509360.1 | 7 | 1026 | |||
XM_011511060.2 | XP_011509362.1 | 6 | 874 | |||
XM_017003940.1 | XP_016859429.1 | 6 | 684 | |||
XM_017003941.1 | XP_016859430.1 | 5 | 801 | |||
XM_017003942.1 | XP_016859431.1 | 5 | 684 | |||
GenBank mRNA | AK296684.1 | - | 5 | 480 | ||
AK300312.1 | - | 6 | 735 | |||
AK312335.1 | - | 5 | 636 | |||
BC001179.1 | - | 5 | 632 | 3528704 | ||
BP283713.1 | - | 5 | 582 | |||
BX647759.1 | - | 5 | 667 | |||
M64098.1 | - | 5 | 585 |