Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005347.4 | NP_005338.1 | 3 | 736 | ||
GenBank mRNA | AF188611.1 | - | 3 | 421 | ||
AF216292.1 | - | 3 | 475 | |||
AJ271729.1 | - | 3 | 488 | |||
AK293604.1 | - | 3 | 692 | |||
BC020235.1 | - | 3 | 679 | 5020098 | ||
EU794617.1 | - | 3 | 488 | |||
X87949.1 | - | 3 | 697 |