Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 3, 4 | 326 | ||
NM_015874.4 | NP_056958.3 | 2, 3 | 273 | |||
NM_203283.2 | NP_976028.1 | 2, 3 | 210 | |||
NM_203284.2 | NP_976029.1 | 3, 4 | 456 | |||
XM_005248161.3 | XP_005248218.1 | 3, 4 | 340 | |||
XM_011513840.2 | XP_011512142.1 | 3, 4 | 732 | |||
XM_017008170.1 | XP_016863659.1 | 2, 3 | 129 | |||
XM_017008171.1 | XP_016863660.1 | 2, 3 | 108 | |||
XM_017008172.1 | XP_016863661.1 | 3, 4 | 271 | |||
XM_017008173.1 | XP_016863662.1 | 3, 4 | 549 | |||
XM_017008174.1 | XP_016863663.1 | 3, 4 | 365 | |||
XM_017008175.1 | XP_016863664.1 | 3, 4 | 281 | |||
GenBank mRNA | AK302230.1 | - | 2, 3 | 63 | ||
AK303244.1 | - | 2, 3 | 122 | |||
AK311419.1 | - | 2, 3 | 109 | |||
BC020780.1 | - | 2, 3 | 210 | 4718839 | ||
BC064976.1 | - | 3, 4 | 231 | 6152924 | ||
BE269143.1 | - | 3, 4 | 236 | |||
BM472788.1 | - | 280 | ||||
BM555733.1 | - | 3, 4 | 434 | |||
BQ216033.1 | - | 2, 3 | 254 | |||
DC361185.1 | - | 2, 3 | 273 | |||
DC389691.1 | - | 3, 4 | 456 | |||
L07872.1 | - | 2, 3 | 168 | |||
L07874.1 | - | 2, 3 | 285 | |||
L07875.1 | - | 3, 4 | 348 | |||
L07876.1 | - | 2, 3 | 213 | |||
Z36843.1 | - | 2, 3 | 211 |