Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14526
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
20, 21
3104
NM_001284215.1
NP_001271144.1
18, 19
2929
NM_001284216.1
NP_001271145.1
17, 18
2150
NM_004238.2
NP_004229.1
19, 20
2960
XM_005246954.4
XP_005247011.1
20, 21
3212
XM_005246955.4
XP_005247012.1
20, 21
3211
XM_005246956.4
XP_005247013.1
20, 21
3188
XM_005246957.4
XP_005247014.1
20, 21
3178
XM_005246958.4
XP_005247015.1
20, 21
3131
XM_005246960.4
XP_005247017.1
21, 22
3125
XM_005246961.3
XP_005247018.1
19, 20
3062
XM_005246962.4
XP_005247019.1
19, 20
3059
XM_006712852.3
XP_006712915.1
19, 20
3015
XM_011512180.2
XP_011510482.1
20, 21
3165
XM_011512181.2
XP_011510483.1
20, 21
3163
XM_011512183.2
XP_011510485.1
19, 20
3019
XM_017005272.1
XP_016860761.1
21, 22
3271
XM_017005273.1
XP_016860762.1
20, 21
3138
XM_017005274.1
XP_016860763.1
20, 21
3086
XM_017005275.1
XP_016860764.1
20, 21
3141
XM_017005276.1
XP_016860765.1
20, 21
3146
XM_017005277.1
XP_016860766.1
21, 22
3286
XM_017005278.1
XP_016860767.1
20, 21
3101
XM_017005279.1
XP_016860768.1
20, 21
3173
XM_017005280.1
XP_016860769.1
20, 21
3162
XM_017005281.1
XP_016860770.1
20, 21
3081
XM_017005282.1
XP_016860771.1
21, 22
3124
XM_017005283.1
XP_016860772.1
20, 21
3128
XM_017005284.1
XP_016860773.1
21, 22
3122
XM_017005285.1
XP_016860774.1
19, 20
3049
XM_017005286.1
XP_016860775.1
19, 20
3081
XM_017005287.1
XP_016860776.1
19, 20
3062
XM_017005288.1
XP_016860777.1
19, 20
3024
XM_017005289.1
XP_016860778.1
21, 22
3044
XM_017005290.1
XP_016860779.1
21, 22
3041
XM_017005291.1
XP_016860780.1
20, 21
2975
XM_017005292.1
XP_016860781.1
19, 20
2978
XM_017005293.1
XP_016860782.1
19, 20
2978
GenBank mRNA
BC113891.1
-
17, 18
2044
40083155
BC114556.1
-
20, 21
2998
40083165
D28476.1
-
19, 20
2890
EU489742.1
-
18, 19
2883
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-4ldn8:80/100.66.77.177:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline