Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 3618 | ||
NM_005512.2 | NP_005503.1 | 3 | 3723 | |||
XM_005273903.4 | XP_005273960.2 | 3 | 3705 | |||
XM_011544935.2 | XP_011543237.1 | 3 | 4014 | |||
GenBank mRNA | BC052210.1 | - | 3 | 3614 | 4825609 | |
BC070079.1 | - | 3 | 3618 | 30344529 | ||
Z24680.1 | - | 3 | 3571 |