Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206567.1 | NP_001193496.1 | 7 | 1608 | ||
NM_005531.2 | NP_005522.2 | 8 | 1776 | |||
XM_005245127.3 | XP_005245184.1 | 9 | 2093 | |||
XM_017001150.1 | XP_016856639.1 | 8 | 1776 | |||
GenBank mRNA | AB208989.1 | - | 8 | 1746 | ||
AF208043.1 | - | 8 | 1750 | |||
AK296228.1 | - | 7 | 1491 | |||
AK307957.1 | - | 8 | 1557 | |||
AY138863.1 | - | 7 | 1486 | |||
BC017059.1 | - | 8 | 1743 | 3914632 | ||
DA945454.1 | - | 2 | 180 | |||
M63838.1 | - | 8 | 1750 |