Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130089.1 | NP_001123561.1 | 6, 7 | 905 | ||
NM_005548.2 | NP_005539.1 | 5, 6 | 725 | |||
XM_017023217.1 | XP_016878706.1 | 5, 6 | 817 | |||
GenBank mRNA | AF285758.1 | - | 6, 7 | 883 | ||
AK126584.1 | - | 4, 5 | 313 | |||
AK309168.1 | - | 5, 6 | 791 | |||
AK315687.1 | - | 5, 6 | 726 | |||
BC004132.2 | - | 5, 6 | 688 | 2963032 | ||
BC013246.1 | - | 5, 6 | 675 | |||
D31890.1 | - | 5, 6 | 691 | |||
D32053.1 | - | 5, 6 | 717 |