Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130089.1 | NP_001123561.1 | 8 | 1100 | ||
NM_005548.2 | NP_005539.1 | 7 | 920 | |||
XM_017023217.1 | XP_016878706.1 | 7 | 1012 | |||
GenBank mRNA | AF285758.1 | - | 8 | 1078 | ||
AK126584.1 | - | 6 | 508 | |||
AK309168.1 | - | 7 | 986 | |||
AK315687.1 | - | 7 | 921 | |||
BC004132.2 | - | 7 | 883 | 2963032 | ||
BC013246.1 | - | 7 | 870 | |||
D31890.1 | - | 7 | 886 | |||
D32053.1 | - | 7 | 912 |