Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201362.1 | NP_001188291.1 | 5 | 1163 | ||
NM_001201363.1 | NP_001188292.1 | 7 | 707 | |||
NM_001318884.1 | NP_001305813.1 | 7 | 714 | |||
NM_001318885.1 | NP_001305814.1 | 5 | 563 | |||
NM_001318886.1 | NP_001305815.1 | 7 | 701 | |||
NM_001318887.1 | NP_001305816.1 | 6 | 1043 | |||
NM_001318888.1 | NP_001305817.1 | 5 | 500 | |||
NM_004242.3 | NP_004233.1 | 6 | 614 | |||
NM_138730.2 | NP_620058.1 | 6 | 573 | |||
GenBank mRNA | AF274949.1 | - | 6 | 533 | ||
AF401520.1 | - | 6 | 464 | |||
AK124181.1 | - | 1 | 1503 | |||
AY043282.1 | - | 6 | 581 | |||
BC009529.2 | - | 6 | 501 | 3890883 | ||
BM927225.1 | - | 7 | 577 | |||
BX648085.1 | - | 1 | 4404 | |||
CF553005.1 | - | 7 | 578 | |||
CK023734.1 | - | 5 | 438 |