Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172557.1 | NP_001166028.1 | 7 | 1770 | ||
NM_005895.3 | NP_005886.2 | 7 | 2108 | |||
XM_005266164.4 | XP_005266221.1 | 7 | 1715 | |||
XM_005266167.4 | XP_005266224.1 | 4 | 1140 | |||
XM_006719736.3 | XP_006719799.1 | 7 | 1835 | |||
XM_006719737.3 | XP_006719800.1 | 7 | 1845 | |||
XM_011534793.2 | XP_011533095.1 | 6 | 1562 | |||
GenBank mRNA | AB027133.1 | - | 7 | 1830 | ||
AF485338.1 | - | 6 | 1552 | |||
BC060826.1 | - | 7 | 1794 | |||
BC142658.1 | - | 7 | 1770 | 40148073 | ||
BC146675.1 | - | 7 | 1770 | 40148852 | ||
D63997.1 | - | 7 | 1818 |