Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006143.2 | NP_006134.1 | 4 | 811 | ||
XM_011520623.2 | XP_011518925.1 | 4 | 1312 | |||
XM_011520624.2 | XP_011518926.1 | 4 | 966 | |||
XM_011520625.2 | XP_011518927.1 | 5 | 1934 | |||
XM_017019212.1 | XP_016874701.1 | 4 | 1824 | |||
XM_017019213.1 | XP_016874702.1 | 4 | 2020 | |||
XM_017019214.1 | XP_016874703.1 | 5 | 2177 | |||
XM_017019215.1 | XP_016874704.1 | 2 | 3154 | |||
XM_017019216.1 | XP_016874705.1 | 4 | 1283 | |||
GenBank mRNA | AK074729.1 | - | 2 | 649 | ||
BC150599.1 | - | 1 | 505 | 9056969 | ||
CR456832.1 | - | 1 | 456 | |||
JQ308175.1 | - | 1 | 456 |
Set Membership: |
G-protein coupled receptor |