Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300914.1 | NP_001287843.1 | 2 | 766 | ||
NM_001313964.1 | NP_001300893.1 | 3 | 692 | |||
NM_006145.2 | NP_006136.1 | 2 | 915 | |||
XM_006722733.1 | XP_006722796.1 | 3 | 705 | |||
XM_006722734.3 | XP_006722797.1 | 3 | 1129 | |||
XM_011527956.2 | XP_011526258.1 | 3 | 684 | |||
GenBank mRNA | AK301817.1 | - | 3 | 607 | ||
AK312624.1 | - | 2 | 776 | |||
BC002352.2 | - | 2 | 739 | 2821008 | ||
BC019827.1 | - | 2 | 729 | 4152608 | ||
CR536485.1 | - | 2 | 705 | |||
CR541677.1 | - | 2 | 705 | |||
D49547.1 | - | 2 | 745 | |||
X62421.1 | - | 2 | 750 |