Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006597.5 | NP_006588.1 | 9 | 2280 | ||
NM_153201.3 | NP_694881.1 | 8 | 1821 | |||
XM_011542798.1 | XP_011541100.1 | 9 | 2028 | |||
GenBank mRNA | AB073886.1 | - | 8 | 1958 | ||
AF217511.1 | - | 3 | 724 | |||
AF352832.1 | - | 9 | 2067 | |||
AK000571.1 | - | 3 | 498 | |||
AK096100.1 | - | 9 | 2006 | |||
AK129885.1 | - | 9 | 2082 | |||
AK222628.1 | - | 8 | 1619 | |||
AK292067.1 | - | 6 | 1710 | |||
BC007276.1 | - | 8 | 1828 | |||
BC008907.2 | - | 4 | 723 | |||
BC015699.1 | - | 4 | 874 | |||
BC016179.1 | - | 9 | 2061 | 3920744 | ||
BC016660.1 | - | 9 | 2061 | 3923981 | ||
BC019816.2 | - | 9 | 2061 | 2899894 | ||
EU668349.1 | - | 9 | 2061 | |||
FJ224294.1 | - | 9 | 2081 |