Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318521.1 | NP_001305450.1 | 6 | 718 | ||
NM_001318523.1 | NP_001305452.1 | 4 | 555 | |||
NM_006733.3 | NP_006724.2 | 5 | 812 | |||
XM_005262111.2 | XP_005262168.1 | 6 | 682 | |||
XM_011530894.2 | XP_011529196.1 | 6 | 711 | |||
XM_011530895.2 | XP_011529197.1 | 7 | 998 | |||
XM_011530897.2 | XP_011529199.1 | 7 | 873 | |||
XM_011530899.2 | XP_011529201.1 | 5 | 679 | |||
XM_017029383.1 | XP_016884872.1 | 6 | 711 | |||
GenBank mRNA | AK302986.1 | - | 6 | 711 | ||
BC012462.2 | - | 4 | 556 | 4537558 | ||
DA736853.1 | - | 6 | 718 | |||
X97249.1 | - | 5 | 811 |