Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006736.5 | NP_006727.2 | 9 | 1164 | ||
GenBank mRNA | AK289626.1 | - | 9 | 987 | ||
AK296297.1 | - | 4 | 714 | |||
AK312723.1 | - | 9 | 1171 | |||
BC011609.2 | - | 9 | 1106 | 3619734 | ||
BT007088.1 | - | 8 | 927 | |||
BX478051.1 | - | 1 | 312 | |||
S37374.1 | - | 8 | 951 |