Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017424.2 | NP_001017424.1 | 7 | 2156 | ||
NM_001017425.2 | NP_001017425.2 | 7 | 1957 | |||
NM_014217.3 | NP_055032.1 | 7 | 1833 | |||
XM_011509522.2 | XP_011507824.1 | 7 | 3359 | |||
XM_011509523.1 | XP_011507825.1 | 7 | 1761 | |||
XM_011509524.2 | XP_011507826.1 | 7 | 1808 | |||
XM_017001248.1 | XP_016856737.1 | 8 | 2544 | |||
XM_017001249.1 | XP_016856738.1 | 8 | 2495 | |||
GenBank mRNA | AK123409.1 | - | 6 | 1727 | ||
AK291483.1 | - | 7 | 1938 |