Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256310.1 | NP_001243239.1 | 2, 3 | 756 | ||
NM_014905.4 | NP_055720.3 | 2, 3 | 756 | |||
XM_005246467.3 | XP_005246524.1 | 2, 3 | 756 | |||
XM_006712435.1 | XP_006712498.1 | 2, 3 | 756 | |||
XM_017003829.1 | XP_016859318.1 | 2, 3 | 756 | |||
GenBank mRNA | AB020645.1 | - | 2, 3 | 742 | ||
AF097492.1 | - | 2, 3 | 390 | |||
AF158555.1 | - | 3, 4 | 736 | |||
AF223943.1 | - | 2, 3 | 511 | |||
AF327434.1 | - | 2, 3 | 722 | |||
AK289747.1 | - | 3, 4 | 732 | |||
BC038507.1 | - | 3, 4 | 732 | 5263220 |