Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256310.1 | NP_001243239.1 | 12 | 1626 | ||
NM_014905.4 | NP_055720.3 | 12 | 1626 | |||
XM_005246467.3 | XP_005246524.1 | 12 | 1626 | |||
XM_006712435.1 | XP_006712498.1 | 12 | 1626 | |||
GenBank mRNA | AB020645.1 | - | 12 | 1612 | ||
AF097492.1 | - | 12 | 1260 | |||
AF097493.1 | - | 5 | 354 | |||
AF158555.1 | - | 13 | 1606 | |||
AF223943.1 | - | 12 | 1381 | |||
AF279697.1 | - | 9 | 711 | |||
AF327434.1 | - | 12 | 1592 | |||
AK001220.1 | - | 9 | 698 | |||
AK289747.1 | - | 13 | 1602 | |||
AK294927.1 | - | 1 | 150 | |||
BC038507.1 | - | 13 | 1602 | 5263220 | ||
CR749593.1 | - | 5 | 2085 |