Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278940.1 | NP_001265869.1 | 10 | 1789 | ||
NM_015859.3 | NP_056943.1 | 9 | 1722 | |||
NM_201595.2 | NP_963889.1 | 9 | 1342 | |||
GenBank mRNA | BX493028.1 | - | 3 | 400 |