Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016602.2 | NP_057686.2 | 2 | 1184 | ||
GenBank mRNA | AF215981.1 | - | 2 | 1184 | ||
BC098132.1 | - | 2 | 1184 | 40004874 | ||
BC098157.1 | - | 2 | 1184 | 40004879 | ||
BC098304.1 | - | 2 | 1184 | 40004880 | ||
BC099840.1 | - | 2 | 1184 | 40004873 | ||
BM930170.1 | - | 1 | 538 |