Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 8 | 763 | ||
NM_001289068.1 | NP_001275997.1 | 8 | 718 | |||
NM_001289069.1 | NP_001275998.1 | 8 | 763 | |||
NM_001289070.1 | NP_001275999.1 | 8 | 763 | |||
NM_001289071.1 | NP_001276000.1 | 9 | 807 | |||
NM_001289072.1 | NP_001276001.1 | 8 | 763 | |||
NM_001289073.1 | NP_001276002.1 | 7 | 698 | |||
NM_001289074.1 | NP_001276003.1 | 8 | 763 | |||
NM_001289075.1 | NP_001276004.1 | 8 | 763 | |||
NM_018063.4 | NP_060533.2 | 8 | 763 | |||
GenBank mRNA | AB102716.1 | - | 9 | 652 | ||
AB102717.1 | - | 7 | 543 | |||
AB102718.1 | - | 8 | 608 | |||
AB102719.1 | - | 8 | 608 | |||
AB102720.1 | - | 8 | 608 | |||
AB102721.1 | - | 8 | 608 | |||
AB102722.1 | - | 8 | 608 | |||
AB102723.1 | - | 8 | 608 | |||
AB113248.1 | - | 8 | 608 | |||
AB113249.1 | - | 8 | 608 | |||
AF155827.1 | - | 8 | 713 | |||
AK022928.1 | - | 2 | 2366 | |||
AK308331.1 | - | 8 | 683 | |||
AK314485.1 | - | 8 | 752 | |||
BX538033.1 | - | 8 | 702 |