Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289067.1 | NP_001275996.1 | 10 | 1155 | ||
NM_001289068.1 | NP_001275997.1 | 10 | 1110 | |||
NM_001289070.1 | NP_001275999.1 | 10 | 1155 | |||
NM_001289071.1 | NP_001276000.1 | 11 | 1199 | |||
NM_001289073.1 | NP_001276002.1 | 9 | 1090 | |||
NM_001289075.1 | NP_001276004.1 | 10 | 1155 | |||
NM_018063.4 | NP_060533.2 | 10 | 1155 | |||
GenBank mRNA | AB102716.1 | - | 11 | 1044 | ||
AB102717.1 | - | 9 | 935 | |||
AB102720.1 | - | 10 | 1000 | |||
AB102721.1 | - | 10 | 1000 | |||
AB102723.1 | - | 10 | 1000 | |||
AB113248.1 | - | 10 | 1000 | |||
AB113249.1 | - | 10 | 1000 | |||
AF155827.1 | - | 10 | 1105 | |||
AK314485.1 | - | 10 | 1144 | |||
BX538033.1 | - | 10 | 1094 |