Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319179.1 | NP_001306108.1 | 12 | 2895 | ||
NM_001319180.1 | NP_001306109.1 | 11 | 2951 | |||
NM_018284.2 | NP_060754.2 | 11 | 2809 | |||
NR_135007.1 | 9 | 2315 | ||||
XM_005270743.2 | XP_005270800.1 | 12 | 2885 | |||
XM_006710574.2 | XP_006710637.1 | 11 | 2742 | |||
XM_011541233.1 | XP_011539535.1 | 10 | 2597 | |||
XM_011541234.1 | XP_011539536.1 | 10 | 2580 | |||
XM_017001013.1 | XP_016856502.1 | 10 | 2729 | |||
XM_017001014.1 | XP_016856503.1 | 9 | 2477 | |||
GenBank mRNA | AK309096.1 | - | 10 | 3449 | ||
AL136680.1 | - | 11 | 2708 | |||
BC063819.1 | - | 9 | 2226 | |||
CR936755.1 | - | 11 | 2811 | |||
CR936758.1 | - | 11 | 2750 |