Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002038.3 | NP_002029.3 | 5 | 728 | ||
NM_022872.2 | NP_075010.1 | 5 | 740 | |||
NM_022873.2 | NP_075011.1 | 5 | 752 | |||
GenBank mRNA | AK024814.1 | - | 5 | 707 | ||
BC011601.2 | - | 5 | 680 | 3140035 | ||
BC015603.2 | - | 5 | 683 | 4654295 | ||
X02492.1 | - | 5 | 674 |