Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 3 | 464 | ||
NM_031243.2 | NP_112533.1 | 4 | 500 | |||
XM_005249729.1 | XP_005249786.1 | 4 | 482 | |||
XM_017012109.1 | XP_016867598.1 | 3 | 448 | |||
XM_017012110.1 | XP_016867599.1 | 3 | 448 | |||
GenBank mRNA | AK129839.1 | - | 3 | 418 | ||
AK289429.1 | - | 4 | 453 | |||
BC000506.2 | - | 3 | 395 | 2822109 | ||
BX537494.1 | - | 2 | 3297 | |||
BX641084.1 | - | 2 | 4037 | |||
M29064.1 | - | 4 | 433 | |||
M29065.1 | - | 3 | 403 |