Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003810.1 | NP_001003810.1 | 4 | 925 | ||
NM_002138.3 | NP_002129.2 | 5 | 982 | |||
NM_031369.2 | NP_112737.1 | 4 | 925 | |||
NM_031370.2 | NP_112738.1 | 5 | 982 | |||
GenBank mRNA | AF039575.1 | - | 5 | 640 | ||
AK057836.1 | - | 5 | 943 | |||
AK292707.1 | - | 4 | 920 | |||
AK300149.1 | - | 5 | 814 | |||
AK303552.1 | - | 3 | 751 | |||
AL832723.1 | - | 1 | 1441 | |||
BC002401.1 | - | 5 | 728 | 2822230 | ||
BC023977.2 | - | 5 | 957 | 2961272 | ||
BC026015.2 | - | 5 | 957 | 2961272 | ||
D55671.1 | - | 4 | 401 | |||
D55672.1 | - | 5 | 949 | |||
D55673.1 | - | 4 | 392 | |||
D55674.1 | - | 5 | 928 | |||
M94630.1 | - | 5 | 622 | |||
X03910.1 | - | 5 | 635 |