Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000188.2 | NP_000179.2 | 18 | 2987 | ||
NM_001322364.1 | NP_001309293.1 | 20 | 3080 | |||
NM_001322365.1 | NP_001309294.1 | 23 | 3385 | |||
NM_001322366.1 | NP_001309295.1 | 18 | 2878 | |||
NM_001322367.1 | NP_001309296.1 | 17 | 2891 | |||
NM_033496.2 | NP_277031.1 | 18 | 2984 | |||
NM_033497.2 | NP_277032.1 | 21 | 3202 | |||
NM_033498.2 | NP_277033.1 | 21 | 3256 | |||
NM_033500.2 | NP_277035.2 | 22 | 3349 | |||
XM_005269737.1 | XP_005269794.1 | 18 | 3258 | |||
XM_011539732.1 | XP_011538034.1 | 22 | 3256 | |||
GenBank mRNA | AB209526.1 | - | 18 | 2983 | ||
AK128226.1 | - | 23 | 3380 | |||
AK292012.1 | - | 18 | 2987 | |||
BC008730.2 | - | 18 | 2942 | 3163058 | ||
M75126.1 | - | 18 | 2967 | |||
X66957.1 | - | 12 | 2029 |