Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000162.3 | NP_000153.1 | 6 | 1104 | ||
NM_033507.1 | NP_277042.1 | 6 | 805 | |||
NM_033508.1 | NP_277043.1 | 7 | 929 | |||
XM_017011966.1 | XP_016867455.1 | 6 | 1120 | |||
GenBank mRNA | AK122876.1 | - | 6 | 799 | ||
BC001890.1 | - | 6 | 1104 | 3536582 | ||
BT007062.1 | - | 6 | 634 | |||
EU008605.1 | - | 6 | 634 | |||
M69051.1 | - | 7 | 916 | |||
M88011.1 | - | 6 | 963 | |||
M90299.1 | - | 6 | 1092 |