Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172128.1 | NP_001165599.1 | 15, 16 | 3687 | ||
NM_004513.5 | NP_004504.3 | 3, 4 | 1439 | |||
NM_172217.3 | NP_757366.2 | 14, 15 | 3427 | |||
XM_005254342.3 | XP_005254399.1 | 15, 16 | 3724 | |||
XM_005254346.4 | XP_005254403.1 | 3, 4 | 2857 | |||
XM_011521519.2 | XP_011519821.1 | 14, 15 | 3427 | |||
XM_011521520.2 | XP_011519822.1 | 15, 16 | 3702 | |||
XM_017022140.1 | XP_016877629.1 | 14, 15 | 3427 | |||
XM_017022141.1 | XP_016877630.1 | 15, 16 | 3686 | |||
XM_017022142.1 | XP_016877631.1 | 15, 16 | 3702 | |||
XM_017022143.1 | XP_016877632.1 | 14, 15 | 3575 | |||
XM_017022144.1 | XP_016877633.1 | 15, 16 | 3740 | |||
XM_017022146.1 | XP_016877635.1 | 5, 6 | 2009 | |||
GenBank mRNA | AK124565.1 | - | 13, 14 | 3029 | ||
AK126222.1 | - | 4, 5 | 2208 | |||
AK128740.1 | - | 9, 10 | 3296 | |||
AK222512.1 | - | 3, 4 | 1364 | |||
AK303659.1 | - | 5, 6 | 2048 | |||
AL833554.1 | - | 17, 18 | 3455 | |||
AY324389.1 | - | 14, 15 | 3326 | |||
BC050362.1 | - | 3, 4 | 1399 | |||
BC064911.1 | - | 3, 4 | 1324 | |||
BC136660.1 | - | 15, 16 | 3452 | 9020650 | ||
CR749286.1 | - | 16, 17 | 4147 | |||
FJ032370.1 | - | 2, 3 | 1237 | |||
M90391.1 | - | 4, 5 | 1526 | |||
S81601.1 | - | 2, 3 | 503 | |||
U82972.1 | - | 3, 4 | 1403 |