Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172128.1 | NP_001165599.1 | 19 | 4387 | ||
NM_004513.5 | NP_004504.3 | 7 | 2142 | |||
NM_172217.3 | NP_757366.2 | 18 | 4130 | |||
XM_005254342.3 | XP_005254399.1 | 19 | 4427 | |||
XM_005254346.4 | XP_005254403.1 | 7 | 3560 | |||
XM_011521520.2 | XP_011519822.1 | 19 | 4405 | |||
XM_017022140.1 | XP_016877629.1 | 18 | 4624 | |||
XM_017022141.1 | XP_016877630.1 | 19 | 4389 | |||
XM_017022142.1 | XP_016877631.1 | 19 | 4402 | |||
XM_017022143.1 | XP_016877632.1 | 18 | 4278 | |||
XM_017022144.1 | XP_016877633.1 | 19 | 4443 | |||
XM_017022146.1 | XP_016877635.1 | 9 | 2712 | |||
GenBank mRNA | AK124565.1 | - | 17 | 3729 | ||
AK126222.1 | - | 8 | 2908 | |||
AK131530.1 | - | 19 | 4386 | |||
AK222512.1 | - | 7 | 2064 | |||
AK303659.1 | - | 10 | 2668 | |||
AL833554.1 | - | 21 | 4652 | |||
BC136660.1 | - | 19 | 4152 | 9020650 | ||
CR749286.1 | - | 20 | 4850 | |||
M90391.1 | - | 8 | 2229 |