Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 12 | 1416 | ||
NM_001281502.1 | NP_001268431.1 | 12 | 1485 | |||
NM_004265.3 | NP_004256.1 | 12 | 1757 | |||
GenBank mRNA | AF084559.1 | - | 12 | 1549 | ||
AF108658.1 | - | 12 | 1342 | |||
AF126799.1 | - | 12 | 1412 | |||
AK027459.1 | - | 6 | 552 | |||
AK027577.1 | - | 6 | 596 | |||
AK074925.1 | - | 10 | 964 | |||
AK074939.1 | - | 12 | 1489 | |||
AK074991.1 | - | 12 | 1225 | |||
AK123240.1 | - | 7 | 1386 | |||
AK290291.1 | - | 12 | 1363 | |||
AK299762.1 | - | 12 | 1485 | |||
AL050118.1 | - | 10 | 1001 |