Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002020.4 | NP_002011.2 | 3 | 387 | ||
NM_182925.4 | NP_891555.2 | 3 | 387 | |||
XM_011534478.2 | XP_011532780.1 | 3 | 765 | |||
XM_017009263.1 | XP_016864752.1 | 3 | 771 | |||
XM_017009264.1 | XP_016864753.1 | 3 | 771 | |||
XM_017009265.1 | XP_016864754.1 | 3 | 772 | |||
XM_017009266.1 | XP_016864755.1 | 3 | 766 | |||
XM_017009267.1 | XP_016864756.1 | 3 | 766 | |||
XM_017009268.1 | XP_016864757.1 | 3 | 330 | |||
GenBank mRNA | AK291679.1 | - | 3 | 363 | ||
AK309910.1 | - | 3 | 363 | |||
AY233382.1 | - | 3 | 328 | |||
AY233383.1 | - | 3 | 328 | |||
DB455035.2 | - | 3 | 387 | |||
EU826564.1 | - | 3 | 308 | |||
EU826565.1 | - | 3 | 308 | |||
EU826566.1 | - | 3 | 308 | |||
U43143.1 | - | 3 | 338 | |||
X68203.1 | - | 3 | 327 | |||
X69878.1 | - | 3 | 329 |